Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAB21L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598453
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598453 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598453 Supplier Thermo Scientific Supplier No. PA598453
Add to Cart
Add to Cart

MAB21L1 Polyclonal Antibody

MAB21L1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAB21L1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MAB21L1
Gene Accession No. Q13394
Gene Alias AW047968; CAGR1; Mab21; mab-21 like 1; MAB21L1; mab-21-like 1 (C. elegans); mab-21-like protein 1; mb211; Nbla00126; protein mab-21-like 1; Putative nucleotidyltransferase MAB21L1
Gene Symbols MAB21L1
Host Species Rabbit
Immunogen Recombinant Human Putative nucleotidyltransferase MAB21L1 protein (1-359AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4081
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.