Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAB21L2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549377

Catalog No. PA549377


Only null left
Add to Cart

Description

Description

Predicted to react with mouse based on sequence homology.

Required for eye morphogenesis. May promote the survival of proliferating retinal progenitor cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAB21L2
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q8UUZ1
mab21l2
KLH conjugated synthetic peptide between 185-219 amino acids from the Central region of human (DANRE) mab21l2.
400 μL
Primary
Zebrafish
Antibody
IgG
Western Blot
Unconjugated
mab21l2
id:ibd3414; LOW QUALITY PROTEIN: protein mab-21-like 2; mab-21 like 2; Mab21L2; mab-21-like 2; mab-21-like 2 (C. elegans); MCOPS14; protein mab-21-like 2; unm_au10
Rabbit
Antigen affinity chromatography, Protein A
RUO
117234
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.