Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAB21L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113545
Description
MAB21L2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Required for eye morphogenesis. May promote the survival of proliferating retinal progenitor cells.
Specifications
| MAB21L2 | |
| Polyclonal | |
| Unconjugated | |
| mab21l2 | |
| id:ibd3414; LOW QUALITY PROTEIN: protein mab-21-like 2; mab-21 like 2; Mab21L2; mab-21-like 2; mab-21-like 2 (C. elegans); MCOPS14; protein mab-21-like 2; unm_au10 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10586, 23937 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.0 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8BPP1, Q9Y586 | |
| mab21l2 | |
| Recombinant Human Protein mab-21-like 2 protein (1-300AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction