Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAB21L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726754 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6754 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6754 Supplier Proteintech Group Inc Supplier No. 109711AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is supposed to recognize both MAB21L2 and its homolog MAB21L1.

Required for eye morphogenesis. May promote the survival of proliferating retinal progenitor cells.

Specifications

Antigen MAB21L2
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MAB21L2
Gene Accession No. Q8BPP1, Q9Y586
Gene Alias mab 21 like 2 (C. elegans), MAB21L2, Protein mab 21 like 2
Gene Symbols MAB21L2
Host Species Rabbit
Immunogen MAB21L2 Fusion Protein Ag1423
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10586, 23937, 680102
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.