Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAB21L3 Antibody (OTI1F6), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272563PECY7
Description
MAB21L3 Monoclonal specifically detects MAB21L3 in Human samples. It is validated for Flow Cytometry.Specifications
MAB21L3 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
MAB21L3 | |
Full length recombinant protein of human MAB21L3 (NP_689580) produced in HEK293T cells. | |
0.1 mL | |
126868.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1F6 | |
Flow Cytometry | |
C1orf161, FLJ38716, mab-21-like 3 (C. elegans), protein mab-21-like 3, RP5-1185H19.1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction