Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MabCaptureC™ High Capacity Protein A Resin

Catalog No. p-9582000
Click to view available options
Quantity:
5 mL
25 mL
100 mL

A high-performance chromatography resin for capturing mAbs and recombinant Fc-tagged fusion proteins. Resin provides high specific binding capacity and enhanced alkaline stability characteristics under clean-in-place conditions (CIP).

MabCaptureC High Capacity Protein A Resin is a high-performance chromatography resin for capturing monoclonal antibodies (mAbs) and recombinant Fc-tagged fusion proteins. It provides high specific-binding capacity and enhanced alkaline-stability characteristics under clean-in-place conditions (CIP), for more robust purification and resin regeneration.

Features of the MabCaptureC High Capacity Protein A Resin include:
High binding capacity—60 mg hIgG/mL at 5-minite residence (80 mg/mL at 10-minute residence)
Alkaline stability—engineered to withstand common CIP procedures with 0.2 M NaOH, enabling efficient cleaning of the chromatography resin and improving product quality
Animal component-free raw materials
Versatility—clean antibody or Fc-fusion protein preparations from cell culture supernatant, serum, or ascites
Stability—high-affinity Protein A is securely immobilized to high purity agarose to prevent leaching of ligand, target antibody, or base support

The Protein A ligand, which is produced in yeast, is immobilized on a rigid and uniformly sized agarose bead with an average particle size of 75 μm. The uniformity of the particle size, in turn, delivers superior performance characteristics over traditional resins.

MabCaptureC High Capacity Protein A Resin, alkaline-stable, is suitable for the purification of mAbs as well as a subpopulation of scFv’s and Fab fragments that contain a VH3 domain.

TRUSTED_SUSTAINABILITY

Specifications

Format Bottle
Binding Capacity 60 mg hIgG/mL @ 5 min residence (80 mg/mL @ 10 min residence)
Support Type Cross linked Agarose
Formulation 50% Slurry
Description High Capacity Protein A Resin, Alkaline Stable
Content And Storage 5 mL of resin provided as a 10 mL 50% slurry. Upon receipt store at 4°C.

Quantity 5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.