Learn More
Thermo Scientific™ MabCaptureC Affinity Matrix

Description
MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast. The engineered rProtein A binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.
MabCaptureC™ Affinity Matrix is suitable for the purification of monoclonal antibodies.The rProtein A ligand is immobilized on a highly cross-linked and uniform-sized agarose-based bead with an average particle size of 75 μm. The uniform particle size delivers superior performance characteristics compared to traditional Protein A resins.
Features of the MabCaptureC™ Affinity Matrix include:
- High dynamic binding capacity for mAbs of >50 g/L at 4.8 min residence time
- Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
- Clean and sanitize with standard sodium hydroxide agents for chromatography products
- Excellent scalability
- Non-animal derived
Main characteristics
- Matrix: agarose-based, epoxide activated
- Average particle size: 75 ± 10 μm
- Ligand: recombinant C domain of Protein A
- Ligand coupling method: epoxide coupling
- Binding capacity: >50 g IgG/L resin at 4.8 min residence time and 10% breakthrough
- Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
- Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
- Operating pressure: ≤3 bar
- Pressure flow: >500 cm/h at 3 bar in a 2.6 x 20 cm column
- Formulation buffer: 20% (v/v) ethanol
For process development and resin screening, MabCaptureC™ is also available in the following formats:
- MabCaptureC™ MiniChrom Column, 1 mL (Cat. No. 5943662001)
- MabCaptureC™ MiniChrom Column for process development, 5 mL (5943662005)
- MabCaptureC™ RoboColumn, 200 μL (5943662200)
- MabCaptureC™ RoboColumn, 600 μL (5943662600)
Specifications
Specifications
| Product Type | Affinity Matrix |
| Binding Capacity | >50 g IgG/L |
| Ligand | Engineered rProtein A |
| Matrix | Agarose-based, epoxide-activated |
| Particle Size | 75 ±10 μm |
| Target | CH2-CH3 interface Fc-region |
| Formulation | Suspension in 20% ethanol |
| For Use With (Application) | Monoclonal antibody purification |
| Quantity | 250 mL |
| Form | Agarose Bead |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.