Learn More
Thermo Scientific™ MabCaptureC™ High Capacity Protein A Resin, 5 mL PROMO
Description
MabCaptureC High Capacity Protein A Resin is a high-performance chromatography resin for capturing monoclonal antibodies (mAbs) and recombinant Fc-tagged fusion proteins. It provides high specific-binding capacity and enhanced alkaline-stability characteristics under clean-in-place conditions (CIP), for more robust purification and resin regeneration.
Features of the MabCaptureC High Capacity Protein A Resin include:
- High binding capacity—60 mg hIgG/mL at 5-minite residence (80 mg/mL at 10-minute residence)
- Alkaline stability—engineered to withstand common CIP procedures with 0.2 M NaOH, enabling efficient cleaning of the chromatography resin and improving product quality
- Animal component-free raw materials
- Versatility—clean antibody or Fc-fusion protein preparations from cell culture supernatant, serum, or ascites
- Stability—high-affinity Protein A is securely immobilized to high purity agarose to prevent leaching of ligand, target antibody, or base support
The Protein A ligand, which is produced in yeast, is immobilized on a rigid and uniformly sized agarose bead with an average particle size of 75 μm. The uniformity of the particle size, in turn, delivers superior performance characteristics over traditional resins.
MabCaptureC High Capacity Protein A Resin, alkaline-stable, is suitable for the purification of mAbs as well as a subpopulation of scFvs and Fab fragments that contain a VH3 domain.
Specifications
Specifications
| Format | Bottle |
| Binding Capacity | 60 mg hIgG/mL @ 5 min residence (80 mg/mL @ 10 min residence) |
| Support Type | Cross linked Agarose |
| Formulation | 50% Slurry |
| Content And Storage | 5 mL of resin provided as a 10 mL 50% slurry. Upon receipt Store at 4°C. |
| Quantity | 5 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.