Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAbPac™ HIC-20 HPLC Columns
GREENER_CHOICE

Catalog No. p-6252903
Change view
Click to view available options
Length (Metric):
10 mm
100 mm
250 mm
Diameter (Metric):
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-898-154 100 mm 4.6 mm
03-898-155 250 mm 4.6 mm
03-898-156 10 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-898-154 Supplier Thermo Scientific™ Supplier No. 088553
Only null left
Add to Cart
Add to Cart

Use this column to separate monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC).

Use the Thermo Scientific™ MAbPac™ HIC-20 HPLC column for separation of monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC). The unique chemistry of this silica column offers high resolution and excellent selectivity. The advanced surface bonding technology provides excellent chemical stability, high resolution, and unique selectivity optimized for mAbs and different from other HIC phases.

Characteristics of the MAbPac HIC-20 Column
This column is based on high-purity, spherical, wide-pore (1,000 Å), 5μm silica particles functionalized with proprietary alkyl amide groups offering:

  • Optimal selectivity for mAbs and related variants
  • Excellent separation of mAb fragments and oxidized variants
  • High resolution and high efficiency
  • Compatible with both organic solvent and aqueous mobile phase
  • Rugged column stability
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Max. Pressure 6000 psi (415 bar)
Particle Size 5 μm
pH 2 to 9
Pore Size 1000 Å
Quantity Each
Temperature 60°C
Stationary Phase Silica
Product Line MAbPac
Packing Material Spherical, High Purity Silica
Column Type Hydrophobic Interaction
Phase Hydrophobic Interaction
Technique Bio-LC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.