Learn More
Description
Separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs) with excellent biocompatibility, low carryover and high resolution.
Use the Thermo Scientific™ MAbPac™ HIC-Butyl HPLC columns to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). This column is based on hydrophilic, nonporous, 5μm polymer particles functionalized with butyl groups. The column's hydrophilic resin provides excellent biocompatibility and low carryover, while the optimal density of the butyl groups provides high resolution separation.
- Optimal selectivity for ADCs
- High resolution and high efficiency
- Stable at wide range of pH (2−12)
- Low carryover
- Rugged column packing
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 100 mm |
| Max. Pressure | 4000 psi (275 bar) |
| Particle Size | 5 μm |
| pH | 2 to 12 |
| Pore Size | Nonporous |
| Quantity | Each |
| Temperature | 60°C |
| Stationary Phase | Silica |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
