Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAbPac™ HIC-Butyl HPLC Columns
GREENER_CHOICE

Catalog No. 03898158 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
100 mm
Diameter (Metric):
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-898-158 10 mm 4.6 mm
03-898-157 100 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-898-158 Supplier Thermo Scientific™ Supplier No. 088559
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs) with excellent biocompatibility, low carryover and high resolution.

Use the Thermo Scientific™ MAbPac™ HIC-Butyl HPLC columns to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). This column is based on hydrophilic, nonporous, 5μm polymer particles functionalized with butyl groups. The column's hydrophilic resin provides excellent biocompatibility and low carryover, while the optimal density of the butyl groups provides high resolution separation.

Features of MabPac HIC-Butyl Columns
  • Optimal selectivity for ADCs
  • High resolution and high efficiency
  • Stable at wide range of pH (2−12)
  • Low carryover
  • Rugged column packing
TRUSTED_SUSTAINABILITY

Specifications

Column Format Guard Column
Diameter (Metric) 4.6 mm
Length (Metric) 10 mm
Max. Pressure 4000 psi (275 bar)
Particle Size 5 μm
pH 2 to 12
Pore Size Nonporous
Quantity Each
Temperature 60°C
Stationary Phase Silica
Product Line MAbPac
Packing Material Hydrophilic Polymer
Column Type Hydrophobic Interaction
Phase Hydrophobic Interaction
Technique Bio-LC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.