Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAbPac™ SCX-10 HPLC Columns
GREENER_CHOICE

Catalog No. 03251006 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
250 mm
Diameter (Metric):
2 mm
4 mm
4.6 mm
9 mm
Particle Size:
5 μm
10 μm
Column Format:
Analytical Column
Guard Column
Preparative Column
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size Column Format
03-251-006 250 mm 4.6 mm 5 μm Analytical Column
DX074625 250 mm 4 mm 10 μm Analytical Column
DX075602 150 mm 4 mm 10 μm Analytical Column
DX075603 50 mm 4 mm 10 μm Analytical Column
DX075604 250 mm 2 mm 10 μm Analytical Column
DXSP6866 250 mm 9 mm 10 μm Preparative Column
03-251-003 150 mm 4 mm 5 μm Analytical Column
03-251-004 50 mm 4.6 mm 5 μm Analytical Column
03-251-005 150 mm 4.6 mm 5 μm Analytical Column
DX078655 250 mm 4 mm 5 μm Analytical Column
DX078656 50 mm 4 mm 5 μm Analytical Column
DX075749 50 mm 2 mm 10 μm Guard Column
DX074631 50 mm 4 mm 10 μm Guard Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 03-251-006 Supplier Thermo Scientific™ Supplier No. 082673
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Use these strong cation-exchange columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants.

Use Thermo Scientific™ MAbPac™ SCX-10 LC Columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.

Superior Performance for mAb Variant Analysis
  • Exceptionally high resolution for mAb variants
  • Ideal for characterization and quality control assessment of mAbs
  • Well-suited for stability studies, mAb therapeutic method development and high-throughput screening
  •  Unmatched column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Meets the regulatory requirements for biopharmaceutical characterization
Multiple Sizes Available
  • Available in 3, 5 and 10μm particle sizes in standard and UHPLC formats
  • Smaller-particle-size columns provide the same resolution as the 10μm columns, but with significantly faster analysis time
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6 mm
Flow Rate 0.8 mL/min
Functionality/Interaction Type Sulfonic
Length (Metric) 250 mm
Max. Pressure 7000 psi (480 bar)
Particle Size 5 μm
pH 2 to 12
Pore Size Nonporous
Quantity Each
Temperature Ambient to 60°C
Stationary Phase Sulfonate
Product Line MAbPac
Packing Material Spherical, Nonporous Polymer
Column Type Ion Exchange
Phase Ion Exchange
Technique Bio-LC
Green Features FSC
Product Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.