Learn More
Description
Use these strong cation-exchange columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants.
Use Thermo Scientific™ MAbPac™ SCX-10 LC Columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.
- Exceptionally high resolution for mAb variants
- Ideal for characterization and quality control assessment of mAbs
- Well-suited for stability studies, mAb therapeutic method development and high-throughput screening
- Unmatched column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
- Meets the regulatory requirements for biopharmaceutical characterization
- Available in 3, 5 and 10μm particle sizes in standard and UHPLC formats
- Smaller-particle-size columns provide the same resolution as the 10μm columns, but with significantly faster analysis time
Specifications
Specifications
| Column Format | Guard Column |
| Diameter (Metric) | 4 mm |
| Length (Metric) | 50 mm |
| Max. Pressure | 3000 psi (207 bar) |
| Particle Size | 10 μm |
| pH | 2 to 12 |
| Pore Size | Nonporous |
| Quantity | Each |
| Temperature | Ambient to 60°C |
| Stationary Phase | Sulfonate |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
