Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva MabSelect Sure™ LX Chromatography Media
GSA_VA

Catalog No. 45002569 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-569 200 mL
45-002-568 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-569 Supplier Cytiva Supplier No. 17547402
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Protein A affinity medium with high dynamic binding capacity which improves performance and cuts costs

Alkali tolerance allows for stability, thorough cleaning and reduction in contamination

  • Alkali-stabilized Protein A-derived (E.coli) Ligand
  • High dynamic binding capacity (DBC) for high titer cultures
  • Reduces risk of contamination and carryover
  • Alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH

Purification of monoclonal antibodies for clinical applications and process scale

Certifications

Certificate of Analysis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
For Use With (Application) For PurifiCatIon of Monoclonal Antibodies for Clinical AppliCatIons
Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
Storage Requirements 2°C to 8°C
Binding Capacity 60 mg Human IgG/mL mediμm
Product Type MabSelect SuRe LX Media
Flow Rate 500 cm/Hoμr
Particle Size 85 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure <2 bar
Matrix Highly Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Alkali-stabilized Protein A-derived (E. coli)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.