Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mac-3 Rat anti-Mouse, FITC, Clone: M3/84, BD

Catalog No. BDB553323 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553323 0.5 mg
BDB562061 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553323 Supplier BD Biosciences Supplier No. 553323
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M3/84 antibody reacts with the 110kDa Mac-3 antigen expressed on mouse mononuclear phagocytes. Surface expression of the 92 to 110kDa glycoprotein antigen increases during differentiation of monocytes to activated peritoneal macrophages. The M3/84 mAb

Host Species: Rat
Clone: M3/84
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse C57Bl/6 peritoneal exudate cells

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mac-3
Applications Flow Cytometry
Classification Monoclonal
Clone M3/84
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse C57Bl/6 peritoneal exudate cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.