Learn More
Invitrogen™ MACC1 Monoclonal Antibody (CL0856)

Description
Immunogen sequence: FRSGRIAQSM SEANLIDMEA GKLSKSCNIT ECQDPDLLHN WPDAFTLRGN NASKVANPFW NQLSASNPF Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 27%.
Specifications
Specifications
| Antigen | MACC1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL0856 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MACC1 |
| Gene Accession No. | Q6ZN28 |
| Gene Alias | 4732474O15Rik; 7A5; Gm267; MACC1; metastasis associated in colon cancer 1; metastasis-associated in colon cancer protein 1; putative binding protein 7a5; RGD1309838; SH3 domain-containing protein 7a5; SH3BP4L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.