Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MACC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556163 Supplier Invitrogen™ Supplier No. PA556163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLLEIMLGNL NTMEALLLEM KIGAEVRKDP FSQVMTEMVC LHSLGKEGPF KVLSNCYIYK DTIQVKLIDL SQVMYLVVAA QAKALPSPAA TIWDYIHKTT SIGIYGPKYI HPSF Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.

MACC1 (Metastasis associated in colon cancer 1) is a key regulator of the hepatocyte growth factor (HGF)-HGF receptor (MET) pathway, which is involved in cellular growth, epithelial-mesenchymal transition, angiogenesis, cell motility, invasiveness, and metastasis. MACC1 protein consists of four domains: ZU5, SH3, and two C-terminal death domains (DD). Expression of MACC1 was found significantly upregulated in malignant tissues (colon cancer of all stages as well as liver and lung metastases) compared to normal tissues or adenomas. MACC1 represents an early and crucial prognostic indicator for colon cancer metastasis that is independent of age, sex, tumor infiltration, nodal status, and lymph vessel invasion. Besides its involvement in signal transduction with the MET receptor, MACC1 also links MET signaling and apoptosis. MACC1 may also be an important therapeutic target for colorectal cancer treatment. At least two isoforms of MACC1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MACC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MACC1
Gene Accession No. Q6ZN28
Gene Alias 4732474O15Rik; 7A5; Gm267; MACC1; metastasis associated in colon cancer 1; metastasis-associated in colon cancer protein 1; putative binding protein 7a5; RGD1309838; SH3 domain-containing protein 7a5; SH3BP4L
Gene Symbols MACC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MACC1. Recombinant protein control fragment (Product #RP-92822).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 346389
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.