Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MACC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578674 Supplier Thermo Scientific Supplier No. PA578674
Add to Cart
Add to Cart

MACC1 Polyclonal Antibody

MACC1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MACC1
Applications Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene MACC1
Gene Accession No. Q6ZN28
Gene Alias 4732474O15Rik; 7A5; Gm267; MACC1; metastasis associated in colon cancer 1; metastasis-associated in colon cancer protein 1; putative binding protein 7a5; RGD1309838; SH3 domain-containing protein 7a5; SH3BP4L
Gene Symbols MACC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human MACC1. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 346389
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.