Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MACF1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB272670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272670 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272670 Supplier Novus Biologicals Supplier No. NBP236528B
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MACF1 Polyclonal antibody specifically detects MACF1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen MACF1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias microtubule-actin crosslinking factor 1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1975 to 2025 of human Microtubule-Actin Crosslinking Factor 1 using the numbering given in entry NP_036222.3.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23499
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.