Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MACF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553081 Supplier Invitrogen™ Supplier No. PA553081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESANVKISGT FSSGWTVRLP EFQFSSQNKE YPDREDCTTE KGKKTTVETE DSSVENPEQD LFVEQKERNP NIDALKVINK VKLEVQRQLI GTQREDQTAV SVRENASRGH LLTIPPAEAE GVPLVVDKDV FSVETPKKE Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 41%.

SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MACF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MACF1
Gene Accession No. Q9UPN3
Gene Alias 620 kDa actin binding protein; 620 kDa actin-binding protein; ABP620; ACF7; Aclp7; Actin cross-linking family 7; actin cross-linking family protein 7; actin-crosslinking protein 7; FLJ45612; FLJ46776; KIAA0465; KIAA0754; KIAA1251; MACF; MACF1; macrophin 1; macrophin 1 isoform; Macrophin-1; microtubule-actin crosslinking factor 1; Microtubule-actin cross-linking factor 1; Microtubule-actin cross-linking factor 1, isoforms 1/2/3/5; mKIAA0465; OFC4; R74989; trabeculin alpha; trabeculin-alpha; Uncharacterized protein KIAA0754
Gene Symbols MACF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MACF1. Recombinant protein control fragment (Product #RP-90749).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23499
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.