Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MACF1 Rabbit anti-Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207651
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7651 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7651 Supplier Fabgennix Inc Supplier No. MACF1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MACF1 Polyclonal antibody specifically detects MACF1 in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen MACF1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene MACF1
Gene Accession No. D3ZHV2, Q9QXZ0
Gene Alias 620 kDa actin binding protein; 620 kDa actin-binding protein; ABP620; ACF7; Aclp7; Actin cross-linking family 7; actin cross-linking family protein 7; actin-crosslinking protein 7; FLJ45612; FLJ46776; KIAA0465; KIAA0754; KIAA1251; MACF; MACF1; macrophin 1; macrophin 1 isoform; Macrophin-1; microtubule-actin crosslinking factor 1; Microtubule-actin cross-linking factor 1; Microtubule-actin cross-linking factor 1, isoforms 1/2/3/5; mKIAA0465; OFC4; R74989; trabeculin alpha; trabeculin-alpha; Uncharacterized protein KIAA0754
Host Species Rabbit
Immunogen Recombinant protein
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11426, 362587
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.