Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Macro H2A.2 Antibody (CL5263), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Macro H2A.2 |
---|---|
Clone | CL5263 |
Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Macro H2A.2 Monoclonal specifically detects Macro H2A.2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Macro H2A.2 | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Unconjugated | |
Mouse | |
core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2 | |
MACROH2A2 | |
IgG1 | |
Protein A purified |
CL5263 | |
Monoclonal | |
Purified | |
Human | |
55506 | |
This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:EKRGRKATSGKKGGK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title