Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Macro H2A.2 Antibody (OTI1C2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272564H
Description
Macro H2A.2 Monoclonal specifically detects Macro H2A.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Macro H2A.2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MACROH2A2 | |
Recombinant protein fragment corresponding to amino acids 2-261 of human Macro H2A.2 (NP_061119) produced in E.coli. | |
0.1 mL | |
55506.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1C2 | |
Western Blot, Immunohistochemistry | |
core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction