Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Macro H2A.2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297224PCP
Description
Macro H2A.2 Polyclonal antibody specifically detects Macro H2A.2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Macro H2A.2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2 | |
Produced in rabbits immunized with E. coli-derived Human Macro H2A.2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55506 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction