Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Macro H2A.2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116340
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116340 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116340 Supplier Invitrogen™ Supplier No. PA5116340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Macro H2A.2 Polyclonal Antibody for Western Blot

Variant histone H2A which replaces conventional H2A in a subset of nucleosomes where it represses transcription. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. May be involved in stable X chromosome inactivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Macro H2A.2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene H2AFY2
Gene Accession No. Q9P0M6
Gene Alias Core histone macro-H2A.2; core histone macroH2A2.2; H2A histone family member Y2; H2A histone family, member Y2; H2AFY2; H2afy3; Histone macroH2A2; histone mH2A2; macroH2A2; mH2A2; RGD1561371
Gene Symbols H2AFY2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 148-169 amino acids from the Central region of human H2AFY2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55506
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.