Learn More
Macro H2A.2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA557437
Description
Variant histone H2A which replaces conventional H2A in a subset of nucleosomes where it represses transcription. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. May be involved in stable X chromosome inactivation.
Immunogen sequence: ILSSKSLVLG QKLSLTQSDI SHIGSMRVEG IVHPTTAEID LKEDIGKALE KAGGKEFLET VKELRKSQGP LHighest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Specifications
Macro H2A.2 | |
Polyclonal | |
Unconjugated | |
H2AFY2 | |
macroH2A2, core histone macro-H2A.2, core histone macroH2A2.2, histone macroH2A2, mH2A2, H2AFY2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
361844, 404634, 55506 | |
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.05mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P0M6, Q8CCK0 | |
H2AFY2 | |
Recombinant protein corresponding to Human Macro H2A.2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only