Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MACROD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567144 Supplier Invitrogen™ Supplier No. PA567144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEIVLATLREW LEQHKDKVDR LIICVFLEKD EDIYRSRLPH YFPVA Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.

Removes ADP-ribose from glutamate residues in proteins bearing a single ADP-ribose moiety. Inactive towards proteins bearing poly-ADP-ribose. Deacetylates O-acetyl-ADP ribose, a signaling molecule generated by the deacetylation of acetylated lysine residues in histones and other proteins. Plays a role in estrogen signaling. Binds to androgen receptor (AR) and amplifies the transactivation function of AR in response to androgen. May play an important role in carcinogenesis and/or progression of hormone-dependent cancers by feed-forward mechanism that activates ESR1 transactivation. Could be an ESR1 coactivator, providing a positive feedback regulatory loop for ESR1 signal transduction. Could be involved in invasive growth by down-regulating CDH1 in endometrial cancer cells. Enhances ESR1-mediated transcription activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MACROD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MACROD1
Gene Accession No. Q9BQ69
Gene Alias [Protein ADP-ribosylaspartate] hydrolase MACROD1; [Protein ADP-ribosylglutamate] hydrolase; [Protein ADP-ribosylglutamate] hydrolase MACROD1; ADP-ribose glycohydrolase MACROD1; ADP-ribose glycohydrolase MACROD1; O-acetyl-ADP-ribose deacetylase MACROD1; AI604841; AW743046; D930010J01Rik; LOC613568 protein; Lrp16; MACRO domain containing 1; MACRO domain-containing protein 1; MACROD1; mono-ADP ribosylhydrolase 1; O-acetyl-ADP-ribose deacetylase MACROD1; O-acetyl-ADP-ribose deacetylase MACROD1; LOW QUALITY PROTEIN: O-acetyl-ADP-ribose deacetylase MACROD1; Protein LRP16
Gene Symbols MACROD1
Host Species Rabbit
Immunogen Recombinant Human MACROD1. Recombinant protein control fragment (Product #RP-107790).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.