Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MACROD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559402
Description
Immunogen sequence: SFLKGLSDKQ REEHYFCKDF VRLKKIPTWK EMAKGVAVKV EEPRYKKDKQ LNEKISLLRS DITKLEV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
Removes ADP-ribose from glutamate residues in proteins bearing a single ADP-ribose moiety. Inactive towards proteins bearing poly-ADP-ribose. Deacetylates O-acetyl-ADP ribose, a signaling molecule generated by the deacetylation of acetylated lysine residues in histones and other proteins. Plays a role in estrogen signaling. Binds to androgen receptor (AR) and amplifies the transactivation function of AR in response to androgen. May play an important role in carcinogenesis and/or progression of hormone-dependent cancers by feed-forward mechanism that activates ESR1 transactivation. Could be an ESR1 coactivator, providing a positive feedback regulatory loop for ESR1 signal transduction. Could be involved in invasive growth by down-regulating CDH1 in endometrial cancer cells. Enhances ESR1-mediated transcription activity.
Specifications
MACROD1 | |
Polyclonal | |
Unconjugated | |
MACROD1 | |
[Protein ADP-ribosylaspartate] hydrolase MACROD1; [Protein ADP-ribosylglutamate] hydrolase; [Protein ADP-ribosylglutamate] hydrolase MACROD1; ADP-ribose glycohydrolase MACROD1; ADP-ribose glycohydrolase MACROD1; O-acetyl-ADP-ribose deacetylase MACROD1; AI604841; AW743046; D930010J01Rik; LOC613568 protein; Lrp16; MACRO domain containing 1; MACRO domain-containing protein 1; MACROD1; mono-ADP ribosylhydrolase 1; O-acetyl-ADP-ribose deacetylase MACROD1; O-acetyl-ADP-ribose deacetylase MACROD1; LOW QUALITY PROTEIN: O-acetyl-ADP-ribose deacetylase MACROD1; Protein LRP16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28992 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BQ69 | |
MACROD1 | |
Recombinant protein corresponding to Human MACROD1. Recombinant protein control fragment (Product #RP-97320). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction