Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Macrophage Antibody (RM0029-11H3), PE/Cy5.5, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212436PECY55
Description
Macrophage Monoclonal specifically detects Macrophage in Mouse samples. It is validated for Flow Cytometry.Specifications
Macrophage | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
Rat | |
Protein G purified | |
Primary | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
RM0029-11H3 | |
Flow Cytometry | |
EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
Isolated mouse macrophages | |
0.1 mL | |
Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction