Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Macrophage and Histiocytoma Marker Antibody (D11), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311429IR
Description
Macrophage and Histiocytoma Marker Monoclonal antibody specifically detects Macrophage and Histiocytoma Marker in Human, Mouse (Negative), Porcine (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Macrophage and Histiocytoma Marker | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Membrane preparation from human hepatocytes | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
D11 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Mouse (Negative), Porcine (Negative), Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction