Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage and Histiocytoma Mouse anti-Human, Clone: D11, Abnova™

Catalog No. 89034753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-753 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-753 Supplier Abnova Corporation Supplier No. MAB14670
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human macrophage and histiocytoma.

  • High specificity
  • High reactivity

Specifications

Antigen Macrophage and Histiocytoma
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone D11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human macrophage and histiocytoma.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS.
Host Species Mouse
Immunogen Membrane preparation from human hepatocytes.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody shows a restricted reactivity to cells of the monocyte/macrophage system and specifically reacts with blood monocytes and stains resident macrophages in a wide variety of human tissues.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.