Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Macrophage Marker Antibody (5/9), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250482AF647
Description
Macrophage Marker Monoclonal specifically detects Macrophage Marker in Human samples. It is validated for Immunohistochemistry.Specifications
Macrophage Marker | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
A cell suspension containing osteoclasts from osteoclastomas. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry | |
5/9 | |
Immunohistochemistry | |
Mouse | |
Protein A purified | |
Immunology | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction