Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage Monoclonal Antibody (MAC387), Abbomax

Catalog No. 502204401
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4401 Supplier Abbomax Inc Supplier No. 605960
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Macrophage Cellular location: Macrophage.

Macrophage Monoclonal antibody specifically detects Macrophage in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Macrophage
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MAC387
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene S100A8
Gene Accession No. P05109
Gene Alias calprotectin; protein S100-A8; S100 calcium binding protein A8; S100A8
Host Species Mouse
Immunogen Enriched cell membrane of Human monocytes
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6279
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.