Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage, Mouse anti-Human, Clone: MAC387, Abnova™

Catalog No. 89999000 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-000 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-000 Supplier Abnova Corporation Supplier No. MAB16142
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen Macrophage
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MAC387
Conjugate Unconjugated
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Host Species Mouse
Immunogen Affinity purified monocyte membrane preparation.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C is stable for two years.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.