Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Macrophages / Histiocytes Monoclonal Antibody (3A5)
GREENER_CHOICE

Catalog No. 01671031
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-031 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-031 Supplier Invitrogen™ Supplier No. MA135593
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody stains macrophages and histocytic cells in a wide variety of normal and diseased tissue. Dendritic cells, myeloid cells and glia cells do not react with this monoclonal antibody. Monocytes stain weakly with 3A5, also after external activation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Macrophages / Histiocytes
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3A5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 1% BSA and 0.1% sodium azide
Gene Alias Histiocyte; Macrophage; Monocyte
Host Species Mouse
Immunogen Proprietary Immunogen.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.