Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Macrophages (Mature) Monoclonal Antibody (25F9)
GREENER_CHOICE

Catalog No. FERMA549061
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549061 Supplier Invitrogen™ Supplier No. MA549061
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody is associated with fully differentiated tissue macrophages both in normal and in diseased tissues, independently of the presence or absence of inflammation. The antigen is absent on freshly isolated monocytes and other blood cells. After 6 to 7 days culture human monocytes become positive. Some melanoma and carcinoma cell lines are also positive. Furthermore the monoclonal antibody 25F9 cross reacts with a subpopulation of macrophages of rhesus monkey, pig alveolar macrophages and Kupffer cells.

Macrophages are a type of white blood cell that plays a crucial role in the immune system. They are responsible for detecting, engulfing, and destroying foreign substances, such as bacteria and viruses, as well as dead cells and debris. Mature macrophages are found in various tissues throughout the body and are highly specialized to perform specific functions depending on their location. For example, alveolar macrophages are found in the lungs and are responsible for removing foreign particles that are inhaled, while Kupffer cells are found in the liver and are responsible for removing old or damaged red blood cells. Understanding the function and behavior of macrophages is crucial for developing new treatments for diseases such as cancer, autoimmune disorders, and infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Macrophages (Mature)
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 25F9
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Accession No. P29466, P48729, Q8N752
Gene Alias Macrophages; Mature Macrophages
Host Species Mouse
Immunogen The monoclonal antibody 25F9 recognizes a protein of 86 kD on the cell surface and within the cytoplasm of mature macrophages.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122011, 1452, 834
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.