Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Macrophages/Monocytes Monoclonal Antibody (KUL01), Biotin
GREENER_CHOICE

Catalog No. PIMA528826
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528826 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528826 Supplier Invitrogen™ Supplier No. MA528826
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Macrophages/Monocytes Monoclonal Antibody for Flow

Macrophages are a large phagocytic cell found in stationary form in the tissues or as a mobile white blood cell, especially at sites of infection. Monocytes are a large phagocytic white blood cell with a simple oval nucleus and clear, grayish cytoplasm. Monocytes and macrophages are found in interdigitating cells and activated microglia cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Macrophages/Monocytes
Applications Flow Cytometry
Classification Monoclonal
Clone KUL01
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.4
Host Species Mouse
Immunogen Chicken peripheral blood mononuclear cell leukocytes.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chicken
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.