Learn More
Invitrogen™ Macrophages/Monocytes Monoclonal Antibody (MOMA-2)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA180630
Description
This clone recognizes an intracellular antigen of mouse macrophages and monocytes. It reacts strongly with macrophages in lymphoid organs such as tingible body macrophages and macrophages in T cell dependant areas. Membrane permeabilization is required for Flow Cytometry.
Macrophages are a large phagocytic cell found in stationary form in the tissues or as a mobile white blood cell, especially at sites of infection. Monocytes are a large phagocytic white blood cell with a simple oval nucleus and clear, grayish cytoplasm. Monocytes and macrophages are found in interdigitating cells and activated microglia cells.
Specifications
Macrophages/Monocytes | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
Rat | |
Protein G | |
RUO | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
MOMA-2 | |
Unconjugated | |
Macrophage; Monocyte | |
Murine lymph node stroma. | |
250 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.