Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mad Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312402
This item is not returnable.
View return policy
Description
Mad Polyclonal antibody specifically detects Mad in Drosophila samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
Mad | |
Polyclonal | |
Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:200 | |
antagonizer of myc transcriptional activity, bHLHc58, MADMAD1, MAX dimerization protein 1, Max dimerizer 1, MAX-binding protein, MGC104659, Protein MAD | |
Synthetic peptide taken within amino acid region 405-455 on Drosophila Mothers against dpp protein. | |
100 μg | |
Signal Transduction | |
4084 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Drosophila | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction