Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MadMax™ Synthetic Leather Palm Mechanics Gloves with TPR Padding

Catalog No. 19824153 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-824-153 X-Large
19-824-151 Medium
19-824-152 Large
19-824-154 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-824-153 Supplier PIP™ Supplier No. 1204000/XL
This item is not returnable. View return policy

Synthetic leather palms enhance grip capabilities in dry or light oil applications, with a PVC diamond grip on the palm and fingertips.

  • Synthetic leather palm provides dexterity, and enhances grip in dry or light oil applications
  • PVC diamond grip on palm and fingertips for extra cut/abrasion resistance
  • Foam laminated spandex back for comfort and breathability
  • Thermoplastic rubber (TPR) molded finger, thumb, and back hand guards for maximum protection
  • Reinforced thumb crotch for extended glove life, enhanced comfort, and increased abrasion resistance
  • Vented lycra fabric keeps hands cool
  • Synthetic leather upper and heavy lycra fabric bottom fourchettes enhance the natural contour of the glove for a better fit
  • Airprene wrist is made with breathable neoprene for comfort
  • Foam padded knuckle protection

Specifications

Back Material TPR Padding
Cuff Material Airprene
Coating Material PVC
Color Hi-vis Yellow
Hand Specific
Material Nylon, Spandex
Product Type Mechanic's Gloves
Size X-Large
Style Gunn
Finger Style Full
For Use With (Application) Construction, Auto Repair, Manufacturing, Mining, and Landscaping
Glove Cuff Color Black
Glove Cuff Style Open
Palm Type Synthetic Leather
Series 120-4000
Thumb Style Wing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.