Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mad Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310418100UL
Description
Mad Polyclonal specifically detects Mad in Mouse samples. It is validated for Western Blot.Specifications
Mad | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
antagonizer of myc transcriptional activity, bHLHc58, MADMAD1, MAX dimerization protein 1, Max dimerizer 1, MAX-binding protein, MGC104659, Protein MAD | |
The immunogen is a synthetic peptide corresponding to a region of Mouse (NP_034881). Peptide sequence TQLKDTECTPVGGPLSLEQVQNGNDTPTQVEYQEAPETQVKARHKEGANQ | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
4084 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction