Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD1L1/MAD1 Antibody (9B10), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1205783FR
Description
MAD1L1/MAD1 Monoclonal specifically detects MAD1L1/MAD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
MAD1L1/MAD1 | |
Monoclonal | |
DyLight 680 | |
hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
MAD1 (9B10) | |
Store at 4C in the dark. |
Immunocytochemistry, Immunofluorescence | |
9B10 | |
Immunocytochemistry/Immunofluorescence | |
MAD1L1 | |
Recombinant full length human MAD1 protein. | |
0.1 mL | |
Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors | |
8379 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction