Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD1L1/MAD1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100569G
Description
MAD1L1/MAD1 Polyclonal antibody specifically detects MAD1L1/MAD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MAD1L1/MAD1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1 | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Mitotic arrest deficient-like 1 using the numbering given in Swiss-Prot entry Q9Y6D9 (GeneID 8379). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors | |
8379 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction