Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD1L1/MAD1 Antibody (SR2235), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322139100UL
Description
MAD1L1/MAD1 Monoclonal antibody specifically detects MAD1L1/MAD1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
MAD1L1/MAD1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR2235 | |
Western Blot 1:500-1:2000 | |
hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1 | |
A synthesized peptide derived from human MAD1L1/MAD1 (Uniprot #: Q9Y6D9) | |
100 μg | |
Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors | |
8379 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction