Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582083 Supplier Invitrogen™ Supplier No. PA582083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NNVVEQLKDW LYKCSVQKLV VVISNIESGE VLERWQFDIE CDKTAKDDSA PREKSQKAIQ DEIRSVIRQI TATVTFLPLL EVSCSFDLLI YTDKDLVVPE KWEESGPQFI TNSEEVRLRS FTTTIH Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 95%.

Tight junction proteins (TJPs) belong to a family of membrane-associated guanylate kinase (MAGUK) homologs that are involved in the organization of epithelial and endothelial intercellular junctions. TJPs bind to the cytoplasmic C termini of junctional transmembrane proteins and link them to the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAD2L1
Gene Accession No. Q13257
Gene Alias AA673185; HsMAD2; J1256; MAD2; MAD2 (mitotic arrest deficient, homolog)-like 1; MAD2 (mitotic arrest deficient, yeast, homolog)-like 1; MAD2 mitotic arrest deficient-like 1; MAD2 mitotic arrest deficient-like 1 (yeast); Mad2a; MAD2L1; MAD2-like 1; MAD2-like protein 1; MadL1; Mitotic arrest deficient 2-like protein 1; mitotic arrest deficient, yeast, homolog-like 1; Mitotic MAD2 protein; Mitotic spindle assembly checkpoint protein MAD2A; Mitotic spindle checkpoint component MAD2; spindle checkpoint protein MAD2; YJL030W
Gene Symbols MAD2L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAD2L1. Recombinant protein control fragment (Product #RP-102062).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.