Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAD2 Recombinant Rabbit Monoclonal Antibody (JU46-13)
MAD2 Recombinant Rabbit Monoclonal Antibody (JU46-13)
Supplier: Thermo Scientific™ MA534624
Description
MAD2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Specifications
MAD2 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q13257 | |
MAD2L1 | |
Recombinant protein within Human Mad2L1 aa 69-124. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
JU46-13 | |
Unconjugated | |
MAD2L1 | |
AA673185; HsMAD2; J1256; MAD2; MAD2 (mitotic arrest deficient, homolog)-like 1; MAD2 (mitotic arrest deficient, yeast, homolog)-like 1; MAD2 mitotic arrest deficient-like 1; MAD2 mitotic arrest deficient-like 1 (yeast); Mad2a; MAD2L1; MAD2-like 1; MAD2-like protein 1; MadL1; Mitotic arrest deficient 2-like protein 1; mitotic arrest deficient, yeast, homolog-like 1; Mitotic MAD2 protein; Mitotic spindle assembly checkpoint protein MAD2A; Mitotic spindle checkpoint component MAD2; spindle checkpoint protein MAD2; YJL030W | |
Rabbit | |
Protein A | |
RUO | |
4085 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction