Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD2L1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100563AF647
Description
MAD2L1 Polyclonal antibody specifically detects MAD2L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MAD2L1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
HSMAD2, MAD2 (mitotic arrest deficient, yeast, homolog)-like 1, MAD2 mitotic arrest deficient-like 1 (yeast), MAD2-like protein 1, MAD2mitotic arrest deficient, yeast, homolog-like 1, Mitotic arrest deficient 2-like protein 1, mitotic spindle assembly checkpoint protein MAD2A | |
The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Mitotic Arrest Deficient 2 using the numbering given in SwissProt entry Q13257 (GeneID 4085). | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
4085 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction