Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD2L1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131311
Description
MAD2L1 Polyclonal specifically detects MAD2L1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
MAD2L1 | |
Polyclonal | |
Unconjugated | |
PBS pH7, 20% Glycerol with 0.025% Proclin 300 | |
MAD2L1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MAD2L1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Expected to cross react based on sequence identity: Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
HSMAD2, MAD2 (mitotic arrest deficient, yeast, homolog)-like 1, MAD2 mitotic arrest deficient-like 1 (yeast), MAD2-like protein 1, MAD2mitotic arrest deficient, yeast, homolog-like 1, Mitotic arrest deficient 2-like protein 1, mitotic spindle assembly checkpoint protein MAD2A | |
Rabbit | |
24 kDa | |
100 μL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
4085 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction