Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD2L1 Monoclonal Antibody (17D10), Active Motif™
SDP

Catalog No. 501992709
Encompass
Change view
Click to view available options
Quantity:
10 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2709 200 μg
50-199-2710 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2709 Supplier Active Motif Supplier No. 39399
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tight junction proteins (TJPs) belong to a family of membrane-associated guanylate kinase (MAGUK) homologs that are involved in the organization of epithelial and endothelial intercellular junctions. TJPs bind to the cytoplasmic C termini of junctional transmembrane proteins and link them to the actin cytoskeleton.

Specifications

Antigen MAD2L1
Applications Western Blot
Classification Monoclonal
Clone 17D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene MAD2L1
Gene Accession No. Q13257, Q9Z1B5
Gene Alias AA673185; HsMAD2; MAD2; MAD2 (mitotic arrest deficient, homolog)-like 1; MAD2 (mitotic arrest deficient, yeast, homolog)-like 1; MAD2 mitotic arrest deficient-like 1; MAD2 mitotic arrest deficient-like 1 (yeast); Mad2a; Mad2l1; MAD2-like 1; MAD2-like protein 1; MadL1; Mitotic arrest deficient 2-like protein 1; mitotic arrest deficient, yeast, homolog-like 1; Mitotic spindle assembly checkpoint protein MAD2A
Gene Symbols MAD2L1
Host Species Mouse
Immunogen This MAD2L1 antibody was raised against full-length recombinant human MAD2L1.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4085, 56150
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.