Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD2L1 Mouse anti-Human, Clone: OTI4D2, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500617

Description
MAD2L1 is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. MAD2L1 is related to the MAD2L2 gene located on chromosome 1. A MAD2 pseudogene has been mapped to chromosome 14.Specifications
MAD2L1 | |
Monoclonal | |
Unconjugated | |
MAD2L1 | |
HSMAD2, MAD2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4085 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4D2 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13257 | |
MAD2L1 | |
Full length human recombit protein of human MAD2L1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction